Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712194.3 | XP_002712240.1 | 4 - 5 | 456 | 71 | |
XM_008258600.2 | XP_008256822.1 | 5 - 6 | 425 | 71 | ||
XM_017342861.1 | XP_017198350.1 | 5 - 6 | 396 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
