Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718891.3 | XP_002718937.1 | 3 - 4 | 596 | 84 | |
XM_008270941.2 | XP_008269163.1 | 3 - 4 | 561 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
