Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718891.3 | XP_002718937.1 | 4 - 5 | 695 | 88 | |
XM_008270941.2 | XP_008269163.1 | 4 - 5 | 660 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
