Assay Details
| Gene Symbol: ALAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720421.3 | XP_002720467.1 | 755 | 56 | ||
| XM_017350076.1 | XP_017205565.1 | 673 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALAD| Gene Symbol: ALAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720421.3 | XP_002720467.1 | 755 | 56 | ||
| XM_017350076.1 | XP_017205565.1 | 673 | 56 | |||
Set Membership: |
|