Assay Details
| Gene Symbol: FCGR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264177.2 | XP_008262399.2 | 1 - 2 | 125 | 105 | |
| XM_008264180.2 | XP_008262402.1 | 1 - 2 | 125 | 105 | ||
More Information
Set Membership: |
|

FCGR2A
Genomic Map