Assay Details
| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708745.3 | XP_002708791.1 | 7 - 8 | 1167 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBRG1| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708745.3 | XP_002708791.1 | 7 - 8 | 1167 | 69 | |
Set Membership: |
|