Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017338802.1 | XP_017194291.1 | 1110 | 65 | ||
XM_017338804.1 | XP_017194293.1 | 1104 | 65 | |||
XM_017338805.1 | XP_017194294.1 | 495 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
