Assay Details
| Gene Symbol: NT5C3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719158.3 | XP_002719204.1 | 8 - 9 | 948 | 71 | |
| XM_008271478.2 | XP_008269700.1 | 7 - 8 | 812 | 71 | ||
More Information
Set Membership: |
|

NT5C3B
Genomic Map