Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719657.3 | XP_002719703.1 | 5 - 6 | 829 | 64 | |
| XM_017349451.1 | XP_017204940.1 | 5 - 6 | 830 | 64 | ||
| XM_017349452.1 | XP_017204941.1 | 5 - 6 | 830 | 64 | ||
| XM_017349453.1 | XP_017204942.1 | 5 - 6 | 830 | 64 | ||
More Information
Set Membership: |
|

ZNF410
Genomic Map