Assay Details
Gene Symbol: FYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713989.3 | XP_002714035.2 | 9 - 10 | 2051 | 72 | |
XM_008262138.2 | XP_008260360.1 | 9 - 10 | 2053 | 72 | ||
XM_008262139.2 | XP_008260361.1 | 9 - 10 | 2002 | 72 | ||
XM_008262140.2 | XP_008260362.1 | 9 - 10 | 1960 | 72 | ||
XM_008262141.2 | XP_008260363.1 | 9 - 10 | 1932 | 72 | ||
XM_008262142.2 | XP_008260364.1 | 9 - 10 | 2038 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
