Assay Details
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008251888.2 | XP_008250110.1 | 234 | 65 | ||
XM_017339520.1 | XP_017195009.1 | 263 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008251888.2, XM_017339520.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
