Assay Details
| Gene Symbol: MTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253518.2 | XP_008251740.1 | 458 | 52 | ||
| XM_008253520.2 | XP_008251742.1 | 718 | 52 | |||
| XM_008253521.2 | XP_008251743.1 | 601 | 52 | |||
| XM_008253522.2 | XP_008251744.1 | 483 | 52 | |||
More Information
Set Membership: |
|

MTG2
Genomic Map