Assay Details
| Gene Symbol: LOC100343106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251255.2 | XP_008249477.1 | 393 | 97 | ||
| XM_017339105.1 | XP_017194594.1 | 508 | 97 | |||
More Information
Set Membership: |
|

LOC100343106
Genomic Map