Assay Details
| Gene Symbol: LOC100344458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709257.3 | XP_002709303.1 | 3 - 4 | 1256 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100344458| Gene Symbol: LOC100344458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709257.3 | XP_002709303.1 | 3 - 4 | 1256 | 118 | |
Set Membership: |
|