Assay Details
Gene Symbol: FOXM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712808.3 | XP_002712854.1 | 7 - 8 | 1484 | 58 | |
XM_002712809.3 | XP_002712855.1 | 8 - 9 | 1529 | 58 | ||
XM_008259662.2 | XP_008257884.1 | 8 - 9 | 1532 | 58 | ||
XM_008259663.2 | XP_008257885.1 | 7 - 8 | 1487 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
