Assay Details
| Gene Symbol: LOC100354908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723116.3 | XP_002723162.1 | 1447 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100354908| Gene Symbol: LOC100354908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723116.3 | XP_002723162.1 | 1447 | 61 | ||
Set Membership: |
|