Assay Details
Gene Symbol: LOC100354908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723116.3 | XP_002723162.1 | 1973 | 90 | ||
XM_017339128.1 | XP_017194617.1 | 1689 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
