Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264166.2 | XP_008262388.1 | 1 - 2 | 579 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP12| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264166.2 | XP_008262388.1 | 1 - 2 | 579 | 60 | |
Set Membership: |
|