Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715392.3 | XP_002715438.2 | 5 - 6 | 893 | 60 | |
| XM_008264585.2 | XP_008262807.1 | 4 - 5 | 737 | 60 | ||
More Information
Set Membership: |
|

HAO2
Genomic Map