Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254148.2 | XP_008252370.1 | 5 - 6 | 668 | 61 | |
| XM_017340272.1 | XP_017195761.1 | 4 - 5 | 578 | 61 | ||
More Information
Set Membership: |
|

CD8B
Genomic Map