Assay Details
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267553.2 | XP_008265775.1 | 9 - 10 | 1819 | 87 | |
XM_008267556.2 | XP_008265778.1 | 7 - 8 | 735 | 87 | ||
XM_017347382.1 | XP_017202871.1 | 8 - 9 | 1725 | 87 | ||
XM_017347383.1 | XP_017202872.1 | 8 - 9 | 1709 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
