Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253873.2 | XP_008252095.1 | 9 | 1643 | 96 | |
| XM_008253883.2 | XP_008252105.1 | 8 | 1549 | 96 | ||
| XM_017340004.1 | XP_017195493.1 | 8 | 1785 | 96 | ||
More Information
Set Membership: |
|

KCNIP3
Genomic Map