Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718925.3 | XP_002718971.1 | 15 - 16 | 2747 | 63 | |
| XM_008271032.2 | XP_008269254.1 | 15 - 16 | 2747 | 63 | ||
More Information
Set Membership: |
|

SPAG5
Genomic Map