Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017348685.1 | XP_017204174.1 | 1 - 2 | 891 | 77 | |
XM_017348686.1 | XP_017204175.1 | 1 - 2 | 894 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017348685.1, XM_017348686.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
