Assay Details
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017343784.1 | XP_017199273.1 | 2 - 3 | 915 | 64 | |
XM_017343786.1 | XP_017199275.1 | 2 - 3 | 248 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
