Assay Details
| Gene Symbol: RNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711071.3 | XP_002711117.1 | 4 - 5 | 771 | 61 | |
| XM_008256645.2 | XP_008254867.1 | 5 - 6 | 865 | 61 | ||
| XM_008256646.2 | XP_008254868.1 | 4 - 5 | 705 | 61 | ||
More Information
Set Membership: |
|

RNF41
Genomic Map