Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341405.1 | XP_017196894.1 | 2 - 3 | 795 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017341405.1 transcripts.
Set Membership: |
|

ELOC
Genomic Map