Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264399.2 | XP_008262621.1 | 1 - 2 | 142 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264399.2 transcripts.
Set Membership: |
|

S100A1
Genomic Map