Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716925.3 | XP_002716971.1 | 7 - 8 | 907 | 51 | |
| XM_017347282.1 | XP_017202771.1 | 8 - 9 | 951 | 51 | ||
More Information
Set Membership: |
|

GLRB
Genomic Map