Assay Details
| Gene Symbol: LOC100349972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251881.2 | XP_008250103.1 | 106 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008251881.2 transcripts.
Set Membership: |
|

LOC100349972
Genomic Map