Assay Details
| Gene Symbol: LOC100349972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251880.2 | XP_008250102.1 | 468 | 76 | ||
| XM_008251881.2 | XP_008250103.1 | 434 | 76 | |||
| XM_017339515.1 | XP_017195004.1 | 608 | 76 | |||
More Information
Set Membership: |
|

LOC100349972
Genomic Map