Assay Details
Gene Symbol: EPB41L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017341647.1 | XP_017197136.1 | 22 | 4533 | 113 | |
XM_017341648.1 | XP_017197137.1 | 22 | 4506 | 113 | ||
XM_017341649.1 | XP_017197138.1 | 21 | 4470 | 113 | ||
XM_017341650.1 | XP_017197139.1 | 21 | 4467 | 113 | ||
XM_017341651.1 | XP_017197140.1 | 20 | 4362 | 113 | ||
XM_017341652.1 | XP_017197141.1 | 21 | 4125 | 113 | ||
XM_017341653.1 | XP_017197142.1 | 21 | 4098 | 113 | ||
XM_017341654.1 | XP_017197143.1 | 20 | 4062 | 113 | ||
XM_017341655.1 | XP_017197144.1 | 20 | 4059 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
