Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262796.2 | XP_008261018.1 | 5 - 6 | 866 | 61 | |
| XM_017345050.1 | XP_017200539.1 | 5 - 6 | 965 | 61 | ||
| XM_017345051.1 | XP_017200540.1 | 5 - 6 | 694 | 61 | ||
| XM_017345052.1 | XP_017200541.1 | 4 - 5 | 611 | 61 | ||
More Information
Set Membership: |
|

PACSIN1
Genomic Map