Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716663.3 | XP_002716709.1 | 12 - 13 | 1640 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP91| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716663.3 | XP_002716709.1 | 12 - 13 | 1640 | 102 | |
Set Membership: |
|