Assay Details
Gene Symbol: LOC100339842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270483.2 | XP_008268705.1 | 5 - 6 | 1835 | 89 | |
XM_008270484.2 | XP_008268706.1 | 5 - 6 | 1832 | 89 | ||
XM_017348549.1 | XP_017204038.1 | 5 - 6 | 666 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
