Assay Details
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339258.1 | XP_017194747.1 | 849 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
