Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711715.3 | XP_002711761.1 | 6 - 7 | 1999 | 64 | |
| XM_017342358.1 | XP_017197847.1 | 6 - 7 | 2095 | 64 | ||
More Information
Set Membership: |
|

ST3GAL2
Genomic Map