Assay Details
Gene Symbol: SYT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723923.3 | XP_002723969.1 | 1066 | 58 | ||
XM_017340037.1 | XP_017195526.1 | 3444 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
