Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264566.2 | XP_008262788.2 | 18 - 19 | 2624 | 93 | |
| XM_008264567.2 | XP_008262789.2 | 18 - 19 | 2623 | 93 | ||
| XM_008264569.2 | XP_008262791.2 | 18 - 19 | 2624 | 93 | ||
| XM_008264570.2 | XP_008262792.2 | 18 - 19 | 2624 | 93 | ||
| XM_008264573.2 | XP_008262795.1 | 18 - 19 | 2678 | 93 | ||
| XM_008264574.2 | XP_008262796.1 | 11 - 12 | 2860 | 93 | ||
| XM_008264575.2 | XP_008262797.1 | 11 - 12 | 1607 | 93 | ||
| XM_008264576.2 | XP_008262798.2 | 18 - 19 | 2623 | 93 | ||
| XM_017345940.1 | XP_017201429.1 | 18 - 19 | 2625 | 93 | ||
| XM_017345941.1 | XP_017201430.1 | 15 - 16 | 2384 | 93 | ||
| XM_017345942.1 | XP_017201431.1 | 14 - 15 | 2022 | 93 | ||
| XM_017345943.1 | XP_017201432.1 | 15 - 16 | 2384 | 93 | ||
| XM_017345944.1 | XP_017201433.1 | 15 - 16 | 2384 | 93 | ||
| XM_017345945.1 | XP_017201434.1 | 18 - 19 | 2625 | 93 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map