Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264566.2 | XP_008262788.2 | 19 - 20 | 2693 | 62 | |
| XM_008264567.2 | XP_008262789.2 | 19 - 20 | 2692 | 62 | ||
| XM_008264569.2 | XP_008262791.2 | 19 - 20 | 2693 | 62 | ||
| XM_008264570.2 | XP_008262792.2 | 19 - 20 | 2693 | 62 | ||
| XM_008264573.2 | XP_008262795.1 | 19 - 20 | 2747 | 62 | ||
| XM_008264574.2 | XP_008262796.1 | 12 - 13 | 2929 | 62 | ||
| XM_008264575.2 | XP_008262797.1 | 12 - 13 | 1676 | 62 | ||
| XM_008264576.2 | XP_008262798.2 | 19 - 20 | 2692 | 62 | ||
| XM_017345940.1 | XP_017201429.1 | 19 - 20 | 2694 | 62 | ||
| XM_017345941.1 | XP_017201430.1 | 16 - 17 | 2453 | 62 | ||
| XM_017345942.1 | XP_017201431.1 | 15 - 16 | 2091 | 62 | ||
| XM_017345943.1 | XP_017201432.1 | 16 - 17 | 2453 | 62 | ||
| XM_017345944.1 | XP_017201433.1 | 16 - 17 | 2453 | 62 | ||
| XM_017345945.1 | XP_017201434.1 | 19 - 20 | 2694 | 62 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map