Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264566.2 | XP_008262788.2 | 29 - 30 | 4359 | 77 | |
| XM_008264567.2 | XP_008262789.2 | 29 - 30 | 4355 | 77 | ||
| XM_008264569.2 | XP_008262791.2 | 29 - 30 | 4359 | 77 | ||
| XM_008264570.2 | XP_008262792.2 | 29 - 30 | 4359 | 77 | ||
| XM_008264573.2 | XP_008262795.1 | 29 - 30 | 4413 | 77 | ||
| XM_008264574.2 | XP_008262796.1 | 22 - 23 | 4595 | 77 | ||
| XM_008264575.2 | XP_008262797.1 | 22 - 23 | 3342 | 77 | ||
| XM_008264576.2 | XP_008262798.2 | 29 - 30 | 4355 | 77 | ||
| XM_017345940.1 | XP_017201429.1 | 29 - 30 | 4360 | 77 | ||
| XM_017345941.1 | XP_017201430.1 | 26 - 27 | 4119 | 77 | ||
| XM_017345942.1 | XP_017201431.1 | 25 - 26 | 3757 | 77 | ||
| XM_017345943.1 | XP_017201432.1 | 26 - 27 | 4119 | 77 | ||
| XM_017345944.1 | XP_017201433.1 | 26 - 27 | 4119 | 77 | ||
| XM_017345945.1 | XP_017201434.1 | 29 - 30 | 4360 | 77 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map