Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264566.2 | XP_008262788.2 | 42 - 43 | 6910 | 102 | |
| XM_008264567.2 | XP_008262789.2 | 42 - 43 | 6906 | 102 | ||
| XM_008264569.2 | XP_008262791.2 | 42 - 43 | 6910 | 102 | ||
| XM_008264570.2 | XP_008262792.2 | 42 - 43 | 6910 | 102 | ||
| XM_008264573.2 | XP_008262795.1 | 42 - 43 | 6964 | 102 | ||
| XM_008264574.2 | XP_008262796.1 | 35 - 36 | 7146 | 102 | ||
| XM_008264575.2 | XP_008262797.1 | 35 - 36 | 5893 | 102 | ||
| XM_008264576.2 | XP_008262798.2 | 42 - 43 | 6906 | 102 | ||
| XM_017345940.1 | XP_017201429.1 | 41 - 42 | 6728 | 102 | ||
| XM_017345941.1 | XP_017201430.1 | 39 - 40 | 6670 | 102 | ||
| XM_017345942.1 | XP_017201431.1 | 38 - 39 | 6308 | 102 | ||
| XM_017345943.1 | XP_017201432.1 | 39 - 40 | 6670 | 102 | ||
| XM_017345944.1 | XP_017201433.1 | 39 - 40 | 6670 | 102 | ||
| XM_017345945.1 | XP_017201434.1 | 41 - 42 | 6728 | 102 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map