Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264570.2 | XP_008262792.2 | 46 - 47 | 7461 | 73 | |
| XM_008264576.2 | XP_008262798.2 | 46 - 47 | 7457 | 73 | ||
| XM_017345944.1 | XP_017201433.1 | 43 - 44 | 7221 | 73 | ||
| XM_017345945.1 | XP_017201434.1 | 45 - 46 | 7279 | 73 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map