Assay Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255683.2 | XP_008253905.1 | 1 - 2 | 145 | 56 | |
| XM_017341410.1 | XP_017196899.1 | 1 - 2 | 414 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008255683.2 transcripts.
Set Membership: |
|

PKIA
Genomic Map