Assay Details
| Gene Symbol: LOC100350957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250673.2 | XP_008248895.1 | 1358 | 69 | ||
| XM_017338750.1 | XP_017194239.1 | 1358 | 69 | |||
More Information
Set Membership: |
|

LOC100350957
Genomic Map