Assay Details
Gene Symbol: LOC100343767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722514.3 | XP_002722560.1 | 1058 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
