Assay Details
| Gene Symbol: LOC103345926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251289.2 | XP_008249511.2 | 419 | 95 | ||
| XM_017339140.1 | XP_017194629.1 | 463 | 95 | |||
More Information
Set Membership: |
|

LOC103345926
Genomic Map