Assay Details
| Gene Symbol: LOC100355864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344286.1 | XP_017199775.1 | 7 - 8 | 837 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100355864| Gene Symbol: LOC100355864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344286.1 | XP_017199775.1 | 7 - 8 | 837 | 109 | |
Set Membership: |
|