Assay Details
| Gene Symbol: LOC103349725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344815.1 | XP_017200304.1 | 1 - 2 | 1556 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103349725| Gene Symbol: LOC103349725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344815.1 | XP_017200304.1 | 1 - 2 | 1556 | 57 | |
Set Membership: |
|