Assay Details
Gene Symbol: LOC4326291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762059.1 | XP_015617545.1 | 2 - 3 | 593 | 71 | |
GenBank mRNA | AK103808.1 | - | 2 - 3 | 600 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
